BAF118470010
Polyethylene carboy with spigot
capacity 4 L (1 gal), polyethylene
Synonyme(s) :
Polyethylene carboy with spigot
199,00 €
About This Item
199,00 €
Produits recommandés
Matériaux
polyethylene
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F11847-0010
Capacité
4 L (1 gal)
Catégories apparentées
Description générale
- Spigot opens fully in one half-turn
- Molded of polyethylene, which resists corrosive acids and alkalis
- Extra heavy walls, yet lighter than glass
Caractéristiques et avantages
- Use to dispense cleaning solutions, disinfectants, laboratory reagents, and intermediate phase mixtures and slurries for various manufacturing processes.
- Lightweight, durable polyethylene with extra-heavy walls for long life and dependability
- Lighter than glass and easy to handle
- Heavy knurling on cap provides excellent grip for opening and closing
- Resistant to corrosive acids and alkalis
- The spigot requires only half turn to open or close
- Smooth sides accept adhesive labels and can be marked with most indelible markers