Skip to Content
Merck
All Photos(2)

Key Documents

810173P

Avanti

16:0-06:0 NBD PA

Avanti Research - A Croda Brand 810173P, powder

Synonym(s):

1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€240.00

€240.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€240.00

About This Item

Empirical Formula (Hill Notation):
C31H54N5O11P
CAS Number:
Molecular Weight:
703.76
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€240.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810173P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810173P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidic acid (PA) is a simple anionic membrane phospholipid.[1] The phospholipid contains phosphomonoester polar head group and two long hydrophobic acyl chains.[2] 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) PA is a fluorescent analog with 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) attached to the sn-2 fatty acid of PA.[3]

Application

16:0-06:0 NBD PA 1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphate is suitable to measure the substrate transport by an increase in cellular fluorescence to determine type 4 P-type ATPases (P4-ATPase) discrimination and its glycerophospholipid (GPL) substrate selection through the directed evolution of a sphingomyelin (SM) flippase.[3]

Biochem/physiol Actions

Phosphatidic acid (PA) is crucial for intracellular signaling involved various cellular processes including membrane trafficking and sperm hyper-activation.[1] It plays a vital role in lipid synthesis.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (810173P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidic acid binding proteins display differential binding as a function of membrane curvature stress and chemical properties
Putta P, et al.
Biochimica et Biophysica Acta, 1858(11), 2709-2716 (2016)
A Chauhan et al.
Neurochemical research, 25(3), 423-429 (2000-04-13)
Fibrillar amyloid beta-protein (Abeta) is the major protein of amyloid plaques in the brains of patients with Alzheimer's disease (AD). The mechanism by which normally produced soluble Abeta gets fibrillized in AD is not clear. We studied the effect of
Priya Putta et al.
Biochimica et biophysica acta, 1858(11), 2709-2716 (2016-08-03)
Phosphatidic acid (PA) is a crucial membrane phospholipid involved in de novo lipid synthesis and numerous intracellular signaling cascades. The signaling function of PA is mediated by peripheral membrane proteins that specifically recognize PA. While numerous PA-binding proteins are known
Mikhail A Zhukovsky et al.
FEBS letters, 593(17), 2428-2451 (2019-08-01)
Phosphatidic acid (PA) is the simplest cellular glycerophospholipid characterized by unique biophysical properties: a small headgroup; negative charge; and a phosphomonoester group. Upon interaction with lysine or arginine, PA charge increases from -1 to -2 and this change stabilizes protein-lipid
Phosphatidic acid in membrane rearrangements
Zhukovsky MA, et al.
Febs Letters, 593(17), 2428-2451 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service