Merck
  • BH3 domain-independent apolipoprotein L1 toxicity rescued by BCL2 prosurvival proteins.
BH3 domain-independent apolipoprotein L1 toxicity rescued by BCL2 prosurvival proteins.
American journal of physiology. Cell physiology (2015-06-26)
J F Heneghan, D H Vandorpe, B E Shmukler, J A Giovinazzo, J A Giovinnazo, J Raper, D J Friedman, M R Pollak, S L Alper
ABSTRACT
The potent trypanolytic properties of human apolipoprotein L1 (APOL1) can be neutralized by the trypanosome variant surface antigen gene product known as serum resistance-associated protein. However, two common APOL1 haplotypes present uniquely in individuals of West African ancestry each encode APOL1 variants resistant to serum resistance-associated protein, and each confers substantial resistance to human African sleeping sickness. In contrast to the dominantly inherited anti-trypanosomal activity of APOL1, recessive inheritance of these two trypanoprotective APOL1 alleles predisposes to kidney disease. Proposed mechanisms of APOL1 toxicity have included BH3 domain-dependent autophagy and/or ion channel activity. We probed these potential mechanisms by expressing APOL1 in Xenopus laevis oocytes. APOL1 expression in oocytes increased ion permeability and caused profound morphological deterioration (toxicity). Coexpression of BCL2 family members rescued APOL1-associated oocyte toxicity in the order MCL1 ∼ BCLW > BCLXL ∼ BCL2A1 ≫ BCL2. Deletion of nine nominal core BH3 domain residues abolished APOL1-associated toxicity, but missense substitution of the same residues abolished neither oocyte toxicity nor its rescue by coexpressed MCL1. The APOL1 BH3 domain was similarly dispensable for the ability of APOL1 to rescue intact mice from lethal trypanosome challenge. Replacement of most extracellular Na(+) by K(+) also reduced APOL1-associated oocyte toxicity, allowing demonstration of APOL1-associated increases in Ca(2+) and Cl(-) fluxes and oocyte ion currents, which were similarly reduced by MCL1 coexpression. Thus APOL1 toxicity in Xenopus oocytes is BH3-independent, but can nonetheless be rescued by some BCL2 family proteins.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
SAFC
HEPES
Sigma-Aldrich
MBCQ, ≥98% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 1.8 gal (6.8 L)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 3.6 gal (13.6 L)
Mini chemical spill kit