Skip to Content
Merck
All Photos(1)

Key Documents

SML0439

Sigma-Aldrich

MBCQ

≥98% (HPLC)

Synonym(s):

4-((3,4-Methylenedioxybenzyl)amino)-6-chloroquinazoline

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€56.30

€56.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€56.30

About This Item

Empirical Formula (Hill Notation):
C16H12ClN3O2
CAS Number:
Molecular Weight:
313.74
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

€56.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Clc1ccc2ncnc(NCc3ccc4OCOc4c3)c2c1

InChI

1S/C16H12ClN3O2/c17-11-2-3-13-12(6-11)16(20-8-19-13)18-7-10-1-4-14-15(5-10)22-9-21-14/h1-6,8H,7,9H2,(H,18,19,20)

InChI key

CNODHOSDWZLJGA-UHFFFAOYSA-N

Biochem/physiol Actions

MBCQ is selective inhibitor of cGMP-specific phosphodiesterase PDE V.
MBCQ is selective inhibitor of cGMP-specific phosphodiesterase PDE V; IC50=19 nM.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masashi Mukohda et al.
The Journal of clinical investigation, 129(6), 2318-2332 (2019-03-22)
Mice selectively expressing PPARγ dominant negative mutation in vascular smooth muscle exhibit RhoBTB1-deficiency and hypertension. Our rationale was to employ genetic complementation to uncover the mechanism of action of RhoBTB1 in vascular smooth muscle. Inducible smooth muscle-specific restoration of RhoBTB1
J F Heneghan et al.
American journal of physiology. Cell physiology, 309(5), C332-C347 (2015-06-26)
The potent trypanolytic properties of human apolipoprotein L1 (APOL1) can be neutralized by the trypanosome variant surface antigen gene product known as serum resistance-associated protein. However, two common APOL1 haplotypes present uniquely in individuals of West African ancestry each encode

Articles

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service