Merck
  • A small chemical library of 2-aminoimidazole derivatives as BACE-1 inhibitors: Structure-based design, synthesis, and biological evaluation.
A small chemical library of 2-aminoimidazole derivatives as BACE-1 inhibitors: Structure-based design, synthesis, and biological evaluation.
European journal of medicinal chemistry (2012-01-03)
Gianpaolo Chiriano, Angela De Simone, Francesca Mancini, Daniel I Perez, Andrea Cavalli, Maria Laura Bolognesi, Giuseppe Legname, Ana Martinez, Vincenza Andrisano, Paolo Carloni, Marinella Roberti
ABSTRACT
In this work, we report a rational structure-based approach aimed at the discovery of new 2-aminoimidazoles as β-secretase inhibitors. Taking advantage of a microwave-assisted synthetic protocol, a small library of derivatives was obtained and biologically evaluated. Two compounds showed promising activities in both enzymatic and cellular assays. Moreover, one of them exhibited the capability to cross the blood-brain barrier as assessed by the parallel artificial membrane permeability assay.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Ofloxacin, VETRANAL®, analytical standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture