Skip to Content
Merck
All Photos(3)

Key Documents

P0847

Sigma-Aldrich

Piroxicam

meets USP testing specifications

Synonym(s):

4-Hydroxy-2-methyl-3-(pyrid-2-yl-carbamoyl)-2H-1,2-benzothiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€104.00
5 G
€358.00
10 G
€664.00

€104.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€104.00
5 G
€358.00
10 G
€664.00

About This Item

Empirical Formula (Hill Notation):
C15H13N3O4S
CAS Number:
Molecular Weight:
331.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

€104.00


Please contact Customer Service for Availability

Request a Bulk Order

Agency

USP/NF
meets USP testing specifications

Quality Level

form

solid

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

CN1C(C(=O)Nc2ccccn2)=C(O)c3ccccc3S1(=O)=O

InChI

1S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19H,1H3,(H,16,17,20)

InChI key

QYSPLQLAKJAUJT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cyclooxygenase inhibitor.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT RE 2 Oral

Target Organs

Gastrointestinal tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andres Lust et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 48(1-2), 47-54 (2012-10-23)
The aim of this study was to gain understanding about the effects of different solid-state forms of a poorly water-soluble piroxicam on drug dissolution and oral bioavailability in rats. Three different solid-state forms of piroxicam were studied: anhydrate I (AH)
D Gerber
Drug intelligence & clinical pharmacy, 21(9), 707-710 (1987-09-01)
During the approximately five years (1981-86) that piroxicam has been available in South Africa, the Medicines Safety Centre has received 31 reports of adverse reactions associated with this drug. Among these are two reactions not previously recorded in the literature
Stéphane Lemaître et al.
Bioorganic & medicinal chemistry, 17(6), 2607-2622 (2009-03-06)
Based on the definition of a 5-HT(4) receptor antagonist pharmacophore, a series of pyrrolo[1,2-a]thieno[3,2-e] and pyrrolo[1,2-a]thieno[2,3-e] pyrazine derivatives were designed, prepared, and evaluated to determine the properties necessary for high-affinity binding to 5-HT(4) receptors. The compounds were synthesized by substituting
K Nezvalová-Henriksen et al.
BJOG : an international journal of obstetrics and gynaecology, 120(8), 948-959 (2013-03-16)
To investigate the individual effects of ibuprofen, diclofenac, naproxen, and piroxicam on pregnancy outcome. Cohort study. Norwegian population. A total of 90 417 women and singleton child pairs. The Norwegian Mother and Child Cohort Study and Medical Birth Registry of
Mariana P Arce et al.
Journal of medicinal chemistry, 52(22), 7249-7257 (2009-10-28)
Novel multifunctional compounds have been designed, synthesized, and evaluated as potential drugs for the treatment of Alzheimer's disease (AD). With an L-glutamic moiety as a suitable biocompatible linker, three pharmacophoric groups were joined: (1) an N-benzylpiperidine fragment selected to inhibit

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service