Skip to Content
Merck
All Photos(1)

Key Documents

V900768

Sigma-Aldrich

Gum arabic from acacia tree

Vetec, reagent grade

Synonym(s):

Acacia gum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201

grade

reagent grade

product line

Vetec

Looking for similar products? Visit Product Comparison Guide

Other Notes

Believed to be a branched polymer of galactose, rhamnose, arabinose, and glucuronic acid as the calcium, magnesium, and potassium salts with a mol. wt. of approx. 250,000.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehrdad Hosseinpour et al.
European journal of pediatric surgery : official journal of Austrian Association of Pediatric Surgery ... [et al] = Zeitschrift fur Kinderchirurgie, 22(3), 234-237 (2012-05-12)
The creation of a stoma on the abdomen and the need for appliance on the peristomal skin make this region vulnerable to inflammatory skin disorders. In this study, we introduce a novel protection with Acacia senegal and compare the protective
Kevin W H Kwok et al.
Aquatic toxicology (Amsterdam, Netherlands), 120-121, 59-66 (2012-05-29)
Silver nanoparticles (AgNPs) with antimicrobial properties are perhaps the most deployed engineered nanomaterials in consumer products. Almost all AgNPs are coated with organic materials to enhance their dispersion in water. Contributions of coatings to the toxicity of NPs have received
Pierre Vanloot et al.
Food chemistry, 135(4), 2554-2560 (2012-09-18)
The authentication of Acacia gums samples requires usually the use of sophisticated and time consuming analytical techniques. There is a need for fast and simple analytical techniques for the objective of a quality control methodology. Commercial Acacia senegal and Acacia
Susana Soares et al.
Journal of agricultural and food chemistry, 60(15), 3966-3972 (2012-03-24)
Condensed tannins are a group of polyphenols that are associated with the astringency sensation, as they readily interact and precipitate salivary proteins. As this interaction is affected by carbohydrates, the aim of this work was to study the effect of
Lynne A Isbell et al.
American journal of physical anthropology, 150(2), 286-300 (2013-01-03)
Patas monkeys (Erythrocebus patas) are midsized primates that feed extensively on the gum of Acacia drepanolobium and the ants are housed in swollen thorns of this Acacia. Their diet resembles that expected more of smaller bodied primates. Patas monkeys are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service