Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8186

Soy Protein Isolate

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€842.00

€842.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€842.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€842.00


Please contact Customer Service for Availability

Request a Bulk Order

packaging

pkg of 50 mg

manufacturer/tradename

NIST®

application(s)

food and beverages

format

mixture

General description

One unit of Standard Reference Material RM8186 comprises 5 packets, with each packet containing around 10 g of powder. For more information, please refer to the COA and SDS.
SRM 8186_cert

SRM 8186_SDS

Application

Reference material (RM) is intended for evaluatingmethods used to determine isoflavones in soy protein isolates and similarmaterials. It is also used for quality assurance when assigning values toin-house reference materials.

Features and Benefits

  • Mass fraction values for isoflavones in soy protein isolate are provided on Dry-Mass Basis and traceable to the measurement processes and standards used by NIST.
  • The non-certified mass fraction values of isoflavones are reported on a dry-mass basis and expressed as mass fraction (mg/kg)
  • Expiration details, storage and use are provided on the NIST certificate.

Other Notes

Reference/Informational value is provided for the following analytes.
Daidzein (C15H10O4), Daidzin (C21H20O9), Genistein (C15H10O5), Genistin (C21H20O10), Glycitein (C16H12O), Glycitin (C22H22O10)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service