Skip to Content
Merck
All Photos(3)

Key Documents

V900010

Sigma-Aldrich

Polyvinylpyrrolidone

Vetec, reagent grade, average mol wt 360,000

Synonym(s):

PVP, Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
12352100

grade

reagent grade

product line

Vetec

mol wt

average mol wt 360,000

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Mei Zhao et al.
Environmental toxicology and chemistry, 32(4), 913-919 (2013-01-25)
The toxicity of manufactured silver nanoparticles (AgNPs) has been widely studied, but the influence of AgNPs on the major ions (such as sodium [Na] and calcium [Ca]) regulations are unknown. In the present study, a freshwater cladoceran Daphnia magna was
X C Qin et al.
Journal of photochemistry and photobiology. B, Biology, 120, 156-162 (2013-01-30)
Nanographene oxide (NGO), a new type of nanomaterial for anticancer drugs delivery and near-infrared (NIR)-mediated photothermal ablation of tumors, has been used in the combination of photothermal therapy and chemotherapy. Herein, targeted chemo-photothermal therapy based on polyvinylpyrrolidone (PVP) functionalized NGO
Markéta Julinová et al.
Water environment research : a research publication of the Water Environment Federation, 84(12), 2123-2132 (2013-01-25)
Polyvinylpyrrolidone (PVP) is a frequently used polymer in the pharmaceutical and foodstuff industries. Because it is not subject to metabolic changes and is virtually nondegradable, trace concentrations of PVP are often found in community wastewaters. The literature finds that the
Sung Neung Lee et al.
Archives of pharmacal research, 36(1), 79-85 (2013-01-19)
A novel surface-attached, spray-dried solid dispersion containing poorly water-soluble carvedilol (CV) without any change in the crystallinity was prepared using water, polyvinylpyrrolidone (PVP K30) and Tween 80. The solid dispersion was optimized by investigating the effects of the weight ratios
Massimiliano di Cagno et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 48(4-5), 775-780 (2013-01-29)
The objective of this study was to investigate if the increase in apparent solubility induced by liposomalization or micellization of the poorly soluble drug hydrocortisone (HC) would lead to an enhancement of its permeability through biological membranes. For this purpose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service