Merck
  • Folic acid-conjugated graphene oxide for cancer targeted chemo-photothermal therapy.
Folic acid-conjugated graphene oxide for cancer targeted chemo-photothermal therapy.
Journal of photochemistry and photobiology. B, Biology (2013-01-30)
X C Qin, Z Y Guo, Z M Liu, W Zhang, M M Wan, B W Yang
ABSTRACT
Nanographene oxide (NGO), a new type of nanomaterial for anticancer drugs delivery and near-infrared (NIR)-mediated photothermal ablation of tumors, has been used in the combination of photothermal therapy and chemotherapy. Herein, targeted chemo-photothermal therapy based on polyvinylpyrrolidone (PVP) functionalized NGO was achieved. Folic acid (FA), a common target molecule to cancer cells, was conjugated to NGO via covalent amide bond. The obtained FA-NGO-PVP was proved to be an ideal pH-responsive nanocarrier for delivery of an anticancer drug doxorubicin (DOX) with the loading ratio more than 100%. In vitro experiments were then performed with the combination of chemotherapy and NIR photothermal therapy. The results demonstrated that the targeted chemo-photothermal therapy could specifically deliver drug and heat to tumor sites and showed excellent efficacy of anticancer therapy. Thus, FA-NGO-PVP could be used as a novel nanomaterial for selective chemo-photothermal therapy.
Sigma-Aldrich
Kollidon® 25
Povidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Povidone, meets USP testing specifications
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000