Skip to Content
Merck
All Photos(1)

Documents

V800362

Sigma-Aldrich

Sodium acetate

suitable for HPLC, ≥99.5%

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

product line

Vetec

Assay

≥99.5%

form

solid

autoignition temp.

1112 °F

technique(s)

HPLC: suitable

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mari Endo et al.
Journal of gastroenterology, 49(6), 1026-1039 (2013-07-13)
Daikenchuto (DKT), a gastrointestinal prokinetic Japanese herbal medicine, is prescribed for patients with postoperative ileus (POI) and adhesive bowel obstruction following abdominal surgery. Several mechanisms for the amelioration of POI by DKT have been suggested; however, it has remained unclear
Carol L Ladner-Keay et al.
PloS one, 9(6), e98753-e98753 (2014-06-04)
The formation of β-sheet rich prion oligomers and fibrils from native prion protein (PrP) is thought to be a key step in the development of prion diseases. Many methods are available to convert recombinant prion protein into β-sheet rich fibrils
Andrew D Shaw et al.
Annals of surgery, 255(5), 821-829 (2012-04-04)
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace
Mayilvahanan Shanmugam et al.
PloS one, 10(2), e0117487-e0117487 (2015-02-24)
Aggregatibacter actinomycetemcomitans a causative agent of periodontal disease in humans, forms biofilm on biotic and abiotic surfaces. A. actinomycetemcomitans biofilm is heterogeneous in nature and is composed of proteins, extracellular DNA and exopolysaccharide. To explore the role played by the
Carola H Blessing et al.
Tree physiology, 35(6), 585-598 (2015-04-17)
The response of carbon allocation to drought has often been studied in terms of short-term transport velocity of recently fixed carbon from leaves to roots and root respiration. However, its dynamic response to other environmental conditions, e.g., to changes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service