Skip to Content
Merck
All Photos(1)

Key Documents

V800245

Sigma-Aldrich

Magnesium sulfate heptahydrate

AR, ≥99.5%

Synonym(s):

Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
form:
crystalline powder

grade

AR

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

Vetec

Assay

≥99.5%

form

crystalline powder

pH

5.0-8.2 (5% in water)

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.001%

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chris Chin
Artificial organs, 37(1), 100-102 (2013-01-12)
Congenital heart disease (CHD) is relatively common and there are treatment options for virtually every form of CHD. With good care, the majority of babies can survive to adulthood; however, CHD remains a significant cause of death in infancy and
A P Morrison et al.
British journal of anaesthesia, 110(5), 702-712 (2013-03-28)
Spinal anaesthesia is the primary anaesthetic technique for many types of surgery. Adjuncts to the local anaesthetics (LA) used in spinal anaesthesia can exhibit undesirable side-effects, limiting their use, but magnesium may have advantages in this respect. We sought randomized
Karen J Gibbins et al.
Obstetrics and gynecology, 121(2 Pt 1), 235-240 (2013-01-25)
Clinical trials support the efficacy and safety of magnesium sulfate for cerebral palsy prevention. We evaluated the implementation of a clinical protocol for the use of magnesium for cerebral palsy prevention in our large women's hospital, focusing on uptake, indications
Jeffrey Michael Smith et al.
BMC pregnancy and childbirth, 13, 34-34 (2013-02-07)
Pre-eclampsia/eclampsia is one of the most common causes of maternal and perinatal morbidity and mortality in low and middle income countries. Magnesium sulfate is the drug of choice for prevention of seizures as part of comprehensive management of the disease.
Christoph Czarnetzki et al.
Anesthesiology, 121(1), 59-67 (2014-03-13)
Magnesium enhances the effect of rocuronium. Sugammadex reverses rocuronium-induced neuromuscular block. The authors investigated whether magnesium decreased the efficacy of sugammadex for the reversal of rocuronium-induced neuromuscular block. Thirty-two male patients were randomized in a double-blinded manner to receive magnesium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service