Documents
BMSW1100200
Sigma-Aldrich
Accuris F1 Grade Calibration Weight
weight set
Synonym(s):
milligram calibration weight, top pan balance weight, top pan calibration
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
manufacturer/tradename
Benchmark Scientific W1100-200
General description
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Precision, compact, and portable balances (readability 0.1 to 0.001g) can be calibrated with our Basic Class F1 Calibration Set. Included in the set are 23 stainless steel cylindrical and leaf weights: 2 x 200g, 1 x 100g, 1 x 50g, 2 x 20g, 1 x 10g, 1 x 5g, 2 x 2g, 1 x 1g, 1 x 500mg, 2 x 200mg, 1 x 100mg, 1 x 50mg, 2 x 20mg, 1 x 10mg, 1 x 5mg, 2 x 2mg, 1 x 1mg. Cylindrical weights are marked with their nominal value. All are supplied in a protective case along with tweezers, a cleaning cloth and dust brush.
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.