Skip to Content
Merck
All Photos(1)

Key Documents

V800130

Sigma-Aldrich

Copper(II) nitrate trihydrate

LR, ≥99%

Synonym(s):

Cupric nitrate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(NO3)2 · 3H2O
CAS Number:
Molecular Weight:
241.60
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99%
grade:
LR
form:
crystalline powder

grade

LR

product line

Vetec

Assay

≥99%

form

crystalline powder

SMILES string

O.O.O.[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Cu.2NO3.3H2O/c;2*2-1(3)4;;;/h;;;3*1H2/q+2;2*-1;;;

InChI key

SXTLQDJHRPXDSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Copper(II) nitrate trihydrate may be used as a copper precursor for the synthesis of a copper(II) complex containing a tridentate (NNS) Schiff base as ligand (HL) that shows moderate antibacterial activity. It is also reported to increase the power conversion efficiency when it is incorporated into titanium dioxide based solar cells followed by treating with sodium hydroxide.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoran Wu et al.
Chemical science, 10(17), 4692-4698 (2019-05-28)
Development of efficient strategies for biomass valorization is a highly attractive topic. Herein, we conducted the first work on electrocatalytic oxidation of renewable furfural to produce the key bioactive intermediate 5-hydroxy-2(5H)-furanone (HFO). It was demonstrated that using H2O as the
Synthesis, crystal structure and antibacterial activities of mixed ligand copper (II) and cobalt (II) complexes of a NNS Schiff base.
Ghosh M, et al.
Polyhedron, 85, 312-319 (2015)
Douglas H Nakahata et al.
Journal of inorganic biochemistry, 187, 85-96 (2018-08-07)
The bis-(1,10-phenanthroline)copper(I) complex, [Cu(I)(phen)2]+, was the first copper-based artificial nuclease reported in the literature. The biological and ligand-like properties of sulfonamides make them good candidates for fine-tuning the reactivity of the [Cu(phen)2] motif with biomolecules. In this context, we developed
Enhancement of TiO 2 Particles Based-Solar Cells Efficiency by Addition of Copper (II) Nitrate and Post-Treatment with Sodium Hydroxyde.
Rokhmat M, et al.
Advanced Materials Research, 1112, 312-319 (2015)
Maxim Zabilskiy et al.
Nature communications, 11(1), 2409-2409 (2020-05-18)
In spite of numerous works in the field of chemical valorization of carbon dioxide into methanol, the nature of high activity of Cu/ZnO catalysts, including the reaction mechanism and the structure of the catalyst active site, remains the subject of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service