Skip to Content
Merck
All Photos(1)

Key Documents

1.02753

Supelco

Copper(II) nitrate trihydrate

for analysis EMSURE®

Synonym(s):

Copper(II) nitrate trihydrate, Copper dinitrate trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€58.80
1 KG
€163.00
25 KG
€2,930.00

€58.80


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€58.80
1 KG
€163.00
25 KG
€2,930.00

About This Item

Linear Formula:
Cu(NO3)2 · 3 H2O
CAS Number:
Molecular Weight:
241.60
MDL number:
UNSPSC Code:
12352302
EC Index Number:
221-838-5
NACRES:
NA.21
form:
solid
solubility:
2670 g/L

€58.80


Estimated to ship on02 April 2025Details


Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

≥99.5% (iodometric)

form

solid

potency

940 mg/kg LD50, oral (Rat)

pH

3-4 (20 °C, 50 g/L in H2O)

mp

114 °C

solubility

2670 g/L

density

2.05 g/cm3 at 20 °C

bulk density

1050 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Fe: ≤0.002%
K: ≤0.01%
Na: ≤0.01%
Ni: ≤0.001%
Pb: ≤0.001%
Zn: ≤0.001%

storage temp.

2-30°C

SMILES string

[Cu+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-]

InChI

1S/Cu.2NO3/c;2*2-1(3)4/q+2;2*-1

InChI key

XTVVROIMIGLXTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (iodometric): ≥ 99.5 %
Chloride (Cl): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.002 %
K (Potassium): ≤ 0.01 %
Na (Sodium): ≤ 0.01 %
Ni (Nickel): ≤ 0.001 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service