Key Documents
Z227285
Supelco® PEEK Fitting
Two-piece with ferrule, fingertight/wrenchtight
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
packaging
pkg of 5 ea
General description
Biocompatible fittings for all 1/16 in. tubing connections.
Features and Benefits
Two-Piece Fingertight/Wrenchtight Fittings: Hold to 6000 psi (420kg/cm2) when finger-tightened or 10,000 psi (700kg/cm2)when wrenchtightened. Double-tapered ferrules included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)