Skip to Content
Merck
All Photos(2)

Key Documents

57654

Upchurch PEEK Fingertight Fitting

Upchurch PEEK Fingertight Fitting

10-32 nut and ferrule

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€90.40

€90.40


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 EA
€90.40

About This Item

UNSPSC Code:
41115700

€90.40


Available to ship on15 April 2025Details


Request a Bulk Order

material

PEEK nut and ferrule

packaging

pkg of 10 ea

manufacturer/tradename

Upchurch Scientific, Inc F300X

I.D.

0.0625 in. (1.5875 mm)

fitting

10-32 nut and ferrule

Looking for similar products? Visit Product Comparison Guide

General description

The one-piece fitting, 55067-U, is convenient to use because the ferrule will not stick in a receiving port and can be easliy found if dropped onto the floor. The two-piece fitting features a separate ferrule so you can replace just the ferrule instead of the entire unit. The Upchurch PEEK Fingertight fittings are for use with 1/16″ O.D. tubing and can be used to 6,000 psi (420 kg/cm2).

Application

Upchurch PEEK fingertight fitting may be used in μ-digestion capillary protease column during two-dimensional (2-D) HPLC analysis.[1] It may also be used as an interface component in capillary electrophoresis inductively coupled plasma mass spectrometer (CE-ICP-MS).[2]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of capillary electrophoresis interfaced to double focusing sector field ICP-MS for nuclide abundance determination of lanthanides produced via spallation reactions in an irradiated tantalum target.
Day, Jason A., et al.
Journal of Analytical Atomic Spectrometry, 15, 1343-1348 (2000)
Characterization of femtomole levels of proteins in solution using rapid proteolysis and nanoelectrospray ionization mass spectrometry.
Blackburn, R. Kevin, and Robert J. Anderegg.
Journal of the American Society For Mass Spectrometry, 8, 483-494 (1997)

Questions

Reviews

Active Filters

  1. Hyderabad
    • Review 1
    • Vote 1
    1 out of 5 stars.

    not satisfied with product quality. material Photo showed online to that of received is different

    The catlog material photo is not matching with that of the received material and product quality is not up to the mark.
    Photo 1 is not matching with photo 2. Seems to be a false claim.
    Not satisfied with product quality.

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for your review.
      The images provided are a representation of the available versions of this product, and may not reflect the specific version you receive. In this case, the second image shows the 'double winged' version, but is not functionally different and should not affect product quality.
      We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service