Skip to Content
Merck
All Photos(1)

Documents

PHL82486

Benzyl cinnamate

phyproof® Reference Substance

Synonym(s):

3-Phenyl 2-propenoic acid benzyl ester, Benzyl 3-phenylpropenoate, Cinnamic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOCH2C6H5
CAS Number:
Molecular Weight:
238.28
Beilstein:
2051339
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

bp

195-200 °C/5 mmHg (lit.)

mp

34-37 °C (lit.)

SMILES string

O=C(OCC1=CC=CC=C1)/C=C/C2=CC=CC=C2

InChI

1S/C16H14O2/c17-16(12-11-14-7-3-1-4-8-14)18-13-15-9-5-2-6-10-15/h1-12H,13H2/b12-11+

InChI key

NGHOLYJTSCBCGC-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Skin Sens. 1B

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

356.0 °F - closed cup

Flash Point(C)

180 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service