Skip to Content
Merck
All Photos(1)

Key Documents

51031

Supelco

Benzyl salicylate

analytical standard

Synonym(s):

Benzyl 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€52.10

€52.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€52.10

About This Item

Linear Formula:
2-(HO)C6H4CO2CH2C6H5
CAS Number:
Molecular Weight:
228.24
Beilstein:
2115365
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€52.10


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.580-1.582
n20/D 1.581 (lit.)

bp

168-170 °C/5 mmHg (lit.)

density

1.176 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Oc1ccccc1C(=O)OCc2ccccc2

InChI

1S/C14H12O3/c15-13-9-5-4-8-12(13)14(16)17-10-11-6-2-1-3-7-11/h1-9,15H,10H2

InChI key

ZCTQGTTXIYCGGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

349.7 °F - closed cup

Flash Point(C)

176.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S362-S380 (2007-11-24)
A toxicologic and dermatologic review of benzyl salicylate when used as a fragrance ingredient is presented.
Marina G Pintado-Herrera et al.
Analytica chimica acta, 851, 1-13 (2014-12-03)
This work presents the development, optimization and validation of a multi-residue method for the simultaneous determination of 102 contaminants, including fragrances, UV filters, repellents, endocrine disruptors, biocides, polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), and several types of pesticides in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service