Skip to Content
Merck
All Photos(1)

Key Documents

ERMBC700

Soya Bean (pesticides)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€610.00

€610.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€610.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€610.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

storage temp.

−20°C

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service