Skip to Content
Merck
All Photos(1)

Key Documents

BCR115

Animal feed (OCP′s)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

30 G
€307.00

€307.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
30 G
€307.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

€307.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

storage temp.

2-8°C

Analysis Note

For more information please see:
BCR115

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants: HCB, β-HCH, γ-HCH, Heptachlor, γ-Chlordane, α-Endosulfan, Dieldrin, Endrin, o,p′-DDT, p,p′-DDE

Matrix Group: Animal Feed

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service