Skip to Content
Merck
All Photos(1)

Key Documents

CRM77069

Supelco

Benzyl salicylate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Benzyl 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€156.00

€156.00


Please contact Customer Service for Availability


Select a Size

Change View
100 MG
€156.00

About This Item

Linear Formula:
2-(HO)C6H4CO2CH2C6H5
CAS Number:
Molecular Weight:
228.24
Beilstein:
2115365
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€156.00


Please contact Customer Service for Availability

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.581 (lit.)

bp

168-170 °C/5 mmHg (lit.)

density

1.176 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

Oc1ccccc1C(=O)OCc2ccccc2

InChI

1S/C14H12O3/c15-13-9-5-4-8-12(13)14(16)17-10-11-6-2-1-3-7-11/h1-9,15H,10H2

InChI key

ZCTQGTTXIYCGGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.
This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

349.7 °F - closed cup

Flash Point(C)

176.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service