Skip to Content
Merck
All Photos(1)

Documents

CRM32967

Supelco

Bispyribac sodium salt

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Sodium 2,6-bis(4,6-dimethoxy-2-pyrimidinyloxy)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N4NaO8
CAS Number:
Molecular Weight:
452.35
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

[Na+].COc1cc(OC)nc(Oc2cccc(Oc3nc(OC)cc(OC)n3)c2C([O-])=O)n1

InChI

1S/C19H18N4O8.Na/c1-26-12-8-13(27-2)21-18(20-12)30-10-6-5-7-11(16(10)17(24)25)31-19-22-14(28-3)9-15(23-19)29-4;/h5-9H,1-4H3,(H,24,25);/q;+1/p-1

InChI key

FUHMZYWBSHTEDZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Application

Bispyribac-sodium is a post-emergence herbicide used in aquatic situations, turf and sports grounds to control grasses, sedges and broad-leaved weeds in paddy rice and other crops. As per the regulation (EC) No 1107/2009 bispyribac-sodium is not approved for use in the European Union as a plant protection product. Maximum residue levels (MRLs) for different products of plant and animal origin have been set according to Reg. (EU) 2019/973 for bispyribac (sum of bispyribac, its salts and its esters, expressed as bispyribac) in the range of 0.01 - 0.05 mg/kg. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.has context menu
Environmental

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service