Skip to Content
Merck
All Photos(1)

Key Documents

32967

Supelco

Bispyribac sodium salt

PESTANAL®, analytical standard

Synonym(s):

Sodium 2,6-bis(4,6-dimethoxy-2-pyrimidinyloxy)benzoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€80.30

€80.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€80.30

About This Item

Empirical Formula (Hill Notation):
C19H17N4NaO8
CAS Number:
Molecular Weight:
452.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€80.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Na+].COc1cc(OC)nc(Oc2cccc(Oc3nc(OC)cc(OC)n3)c2C([O-])=O)n1

InChI

1S/C19H18N4O8.Na/c1-26-12-8-13(27-2)21-18(20-12)30-10-6-5-7-11(16(10)17(24)25)31-19-22-14(28-3)9-15(23-19)29-4;/h5-9H,1-4H3,(H,24,25);/q;+1/p-1

InChI key

FUHMZYWBSHTEDZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Bispyribac sodium belongs to the class of pyrimidinyl carboxy herbicides utilized for the post-emergence control of cyperaceous and dicotyledonous grasses and broad-leaf weeds.[1]

Application

Bispyribac sodium may be used as an analytical reference standard for the determination of the analyte in:
  • Water samples[2] and agricultural products[3][4][5] by liquid chromatography-tandem mass spectrometry (LC-MS/MS).
  • Fruits and vegetables by liquid chromatography–quadrupole-time of flight–mass spectrometry (LC-QTOF-MS) with automated detection.[6]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradation of bispyribac sodium by a novel bacterial consortium BDAM: Optimization of degradation conditions using response surface methodology.
Ahmad F, et al.
Journal of Hazardous Materials, 349, 272-281 (2018)
Evaluation and validation of an accurate mass screening method for the analysis of pesticides in fruits and vegetables using liquid chromatography-quadrupole-time of flight-mass spectrometry with automated detection.
Lopez M, et al.
Journal of Chromatography A, 1373(1), 40-50 (2014)
Evaluation of an alternative fluorinated sorbent for dispersive solid-phase extraction clean-up of the quick, easy, cheap, effective, rugged, and safe method for pesticide residues analysis.
Martins ML, et al.
Journal of Chromatography A, 1514(1), 36-43 (2017)
Multi-residue method for determination of 58 pesticides, pharmaceuticals and personal care products in water using solvent demulsification dispersive liquid-liquid microextraction combined with liquid chromatography-tandem mass spectrometry.
Caldas SS, et al.
Talanta, 146, 676-688 (2016)
Simultaneous determination of herbicides in rice by QuECHERS and LC-MS/MS using matrix-matched calibration.
Rebelo AM, et al.
Journal of the Brazilian Chemical Society, 27(1), 186-193 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service