Skip to Content
Merck
All Photos(1)

Key Documents

94961

Terbuthylazine Metabolite CSAA036479 hydrochloride

PESTANAL®, analytical standard

Synonym(s):

2-Hydroxy-4-(1-carboxy-1-methylethylamino)-6-aminotriazine hydrochloride, 2-[(4-Amino-6-hydroxy-1,3,5-triazin-2-yl)amino]-2-methylpropanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

94961-10MG
€51.00

€51.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
94961-10MG
€51.00

About This Item

Empirical Formula (Hill Notation):
C7H12ClN5O3
Molecular Weight:
249.65
UNSPSC Code:
41116107

€51.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

product line

PESTANAL®

Assay

≥95.0 % (HPLC)

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

OC1=NC(NC(C)(C)C(O)=O)=NC(N)=N1.Cl

InChI

1S/C7H11N5O3.ClH/c1-7(2,3(13)14)12-5-9-4(8)10-6(15)11-5;/h1-2H3,(H,13,14)(H4,8,9,10,11,12,15);1H

InChI key

QQWQWJFOHCPBKX-UHFFFAOYSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service