Skip to Content
Merck
All Photos(2)

Documents

37898

Supelco

Trinexapac-ethyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16O5
CAS Number:
Molecular Weight:
252.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

36-39 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(CC(C(OCC)=O)CC1=O)C1=C(C2CC2)O

InChI

1S/C13H16O5/c1-2-18-13(17)8-5-9(14)11(10(15)6-8)12(16)7-3-4-7/h7-8,16H,2-6H2,1H3/b12-11-

InChI key

RVKCCVTVZORVGD-QXMHVHEDSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Trinexapac-ethyl, a compound that belongs to the cyclohexanedione group, is a growth regulator, which can find applications in agriculture and horticulture. Its mode of action basically involves the suppression of gibberellin biosynthesis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Skin Sens. 1B - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Trinexapac-ethyl influence on cell membrane thermostability of Kentucky bluegrass leaf tissue
Heckman.LN
Sci. Hortic., 92, 183-186 (2002)
Influence of trinexapac-ethyl on growth and development of winter wheat
Matysiak K
Journal of Plant Protection Research, 46, 133-143 (2006)
Chayanis Sutcharitchan et al.
Food chemistry, 322, 126766-126766 (2020-04-20)
A simple, rapid, and effective method based on salting-out extraction and LC-MS/MS techniques was developed for the determination of 39 plant growth regulator (PGR) residues in two of the most common root and rhizome Chinese herbs, Codonopsis Radix (Dangshen) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service