Skip to Content
Merck
All Photos(1)

Key Documents

72663

Supelco

5-Chloro-2-hydroxybenzophenone

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€49.90

€49.90


Please contact Customer Service for Availability


Select a Size

Change View
100 MG
€49.90

About This Item

Linear Formula:
ClC6H3(OH)COC6H5
CAS Number:
Molecular Weight:
232.66
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€49.90


Please contact Customer Service for Availability

grade

analytical standard

Quality Level

Assay

≥95%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

95-98 °C
96-98 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Oc1ccc(Cl)cc1C(=O)c2ccccc2

InChI

1S/C13H9ClO2/c14-10-6-7-12(15)11(8-10)13(16)9-4-2-1-3-5-9/h1-8,15H

InChI key

OMWSZDODENFLSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service