5-Chloro-2-hydroxybenzophenone
Linear Formula:
ClC6H3(OH)COC6H5
CAS Number:
Molecular Weight:
232.66
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing