Skip to Content
Merck
All Photos(1)

Key Documents

59138

Supelco

SUPELCOSIL SAX1 (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOSIL SAX1 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

Agency

suitable for USP L14

manufacturer/tradename

SUPELCOSIL

extent of labeling

12% carbon loading

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

170 m2/g

matrix

silica gel, spherical particle platform

matrix active group

propyltrimethylammonium phase

particle size

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL SAX1 HPLC Column is typically employed as an anion exchange column in chromatographic separations.[1]
The SUPELCOSIL SAX1 column has a strongly basic quaternary aminopropyl phase and is used for separating anions.

Application

SUPELCOSIL SAX1 HPLC Column has been used for the separation of:
  • Organoarsenic compounds in marine algae[1] and in biological materials[2] during their analysis using size-exclusion-anion-exchange (SE-AE) high performance liquid chromatography (HPLC) combined with inductively coupled plasma mass spectrometry (ICP-MS).
  • Organoarsenic compounds in edible algae during their analysis using bi-dimensional SE-AE-HPLC combined with dual ICP-MS and electrospray tandem mass spectrometry (ESI-MS/MS).[3]
  • Polypeptides in biological matrices during their analysis using supercritical fluid chromatography (SFC) coupled to MS detector.[4]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas and liquid chromatography with inductively coupled plasma mass spectrometry detection for environmental speciation analysis-advances and limitations.
Szpunar J, et al.
Spectrochimica Acta. Part B: Atomic Spectroscopy, 55(7), 779-793 (2000)
Speciation of arsenic in edible algae by bi-dimensional size-exclusion anion exchange HPLC with dual ICP-MS and electrospray MS/MS detection.
McSheehy S and Szpunar J
Journal of Analytical Atomic Spectrometry, 15(1), 79-87 (2000)
Feasibility of supercritical fluid chromatography/mass spectrometry of polypeptides with up to 40-mers.
Zheng J, et al.
Analytical Chemistry, 78(5), 1535-1545 (2006)
Trace element speciation analysis of biomaterials by high-performance liquid chromatography with inductively coupled plasma mass spectrometric detection.
Szpunar J.
TrAC, Trends in Analytical Chemistry, 19(2-3), 127-137 (2000)

Protocols

EPA Method 547 outlines the analysis of glyphosate in drinking water by direct aqueous injection HPLC, post column derivatization, and fluorescence detection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service