Skip to Content
Merck
All Photos(1)

Documents

59026

Valco® Unions

internal reducing, for connecting 1/16 to 1/32 in., bore size 0.25 mm

Synonym(s):

1/16" TO 1/32" REDUCING UNION

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

stainless steel union and fittings

packaging

pkg of 1 ea

manufacturer/tradename

VICI Valco® ZRU1.5

bore size

0.25 mm

fitting

10-32 Valco for 1/16 in. O.D. tubing (nuts and ferrules included)
6-32 Valco for 1/32 in. O.D. tubing (nuts and ferrules included)

compatibility

for connecting 1/16 to 1/32 in.

Looking for similar products? Visit Product Comparison Guide

General description

Stainless steel, unless noted otherwise. Nuts and ferrules included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service