Key Documents
58289
Waters® Union
size 1/16 in., stainless steel
Select a Size
1 EA
€98.60
€98.60
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
€98.60
About This Item
UNSPSC Code:
41115700
€98.60
Please contact Customer Service for Availability
Request a Bulk Order
material
stainless steel
packaging
pkg of 1 ea
size
1/16 in.
General description
Waters® Union is used for connecting most HPLC columns to a 1/16 in. tubing system.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Waters is a registered trademark of Waters
Choose from one of the most recent versions:
Certificates of Analysis (COA)