Key Documents
57123
Supelco
SPE Tube Adapter
configured for AutoTrace Automated Systems, for use with 3mL tubes, pk of 6
Select a Size
6 EA
€46.90
€46.90
Please contact Customer Service for Availability
Request a Bulk OrderRequest a Sample
Select a Size
Change View
6 EA
€46.90
About This Item
UNSPSC Code:
12000000
€46.90
Please contact Customer Service for Availability
Request a Bulk OrderRequest a Sample
packaging
pk of 6
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
configured for AutoTrace Automated Systems
for use with 3mL tubes
General description
Tube adapters serve many purposes. They can be used to stack one SPE tube on top of another to provide different selectivities. A larger empty syringe barrel can be stacked on top of a smaller SPE tube to act as a larger load reservoir. Or, they can serve as an adapter for positive pressure methods (e.g. from a syringe or air/ N2 line).
Choose from one of the most recent versions:
Certificates of Analysis (COA)