Skip to Content
Merck
All Photos(1)

Documents

51354-U

MXP High Pressure Valves for HPLC

stainless steel, pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

material

stainless steel

description

MXP7900-000 2-position, 6-port

packaging

pkg of 1 ea

Looking for similar products? Visit Product Comparison Guide

General description

The MXP7900-000 valve is the standard 2-position, 6-port switching valve designed for traditional HPLC and related techniques, used for traditional sample injections or to provide dual-column functionality in your system.

The MXP9900-000 valve should be used when the application requires biocompatability.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service