Key Documents
Z225320
Sigma-Aldrich
Aldrich® SAFE-PURGE valve
manual
Select a Size
1 EA
€577.00
€577.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€577.00
About This Item
UNSPSC Code:
40141600
NACRES:
NB.79
€577.00
Please contact Customer Service for Availability
parameter
<5 psi pressure
General description
The Aldrich SAFE-PURGE valves provide for safe, high-performance operation. Both the manual and automatic spring-loaded styles feature a durable PTFE metering valve.
  • Quick and efficient purging of reaction vessel which conserves expensive purge gases
  • Safely vents toxic gases from built-in oil bubbler
Used with purge gas pressure under 5 psi.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SAFE-PURGE is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)