50660-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column
column L × I.D. 50 mm × 3.0 mm, pk of 1 ea
About This Item
Recommended Products
CofA
current certificate can be downloaded
Quality Level
packaging
pk of 1 ea
extent of labeling
5.1% Carbon loading
parameter
600 bar max. pressure (8700 psi)
maximum temperature
60 °C (pH2/60; pH7/40)
column L × I.D.
50 mm × 3.0 mm
surface area
75 m2/g
matrix
superficially porous particle particle platform (Fused-Core®)
matrix active group
C18 bonding phase, dimethyloctadecylsilane and positively charged ligand
operating pH range
2-7
Looking for similar products? Visit Product Comparison Guide
General description
Analysis Note
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service