64065-U
Ascentis® Express 90 Å Biphenyl (2.7 µm) HPLC Columns
L × I.D. 10 cm × 2.1 mm, HPLC Column
Select a Size
€801.00
Select a Size
About This Item
€801.00
Recommended Products
Product Name
Ascentis® Express Biphenyl Column, 2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
material
stainless steel hardware
Quality Level
Agency
suitable for USP L11
product line
Ascentis®
feature
endcapped
manufacturer/tradename
Ascentis®
packaging
1 ea of
parameter
60 °C max. temp.
9000 psi max. pressure (600 bar)
technique(s)
HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable
L × I.D.
10 cm × 2.1 mm
surface area
135 m2/g
impurities
<5 ppm metals
matrix
particle platform (Fused-Core)
superficially porous particle
matrix active group
biphenyl phase
particle size
2.7 μm
pore size
90 Å pore size
operating pH
2-9
application(s)
food and beverages
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
guard column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Determination of plasma protein binding using a SupelTM C18 pin device based BioSPME method for efficient sample preparation, along with a comparison to the equilibrium dialysis method.
The application note describes a comparative analysis of plasma protein binding results for free/bound drugs using two different sample extraction steps, with and without agitation.
Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.
Learn about Supel™ BioSPME C18 high-throughput devices to determine plasma protein binding and decrease the sample preparation time to less than an hour.
Protocols
HPLC-UV analysis of active compounds in marijuana-infused edibles for potency testing; complex matrix requires precise methods.
Rapid potency testing of marijuana-infused edibles using LC/MS on a biphenyl stationary phase detected eleven cannabinoids.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service