Key Documents
504351
Supelco
Male Luer Plug
configured for plugging luer holes, pkg of 12 ea
Select a Size
12 EA
€34.10
€34.10
Please contact Customer Service for Availability
Request a Bulk OrderRequest a Sample
Select a Size
Change View
12 EA
€34.10
About This Item
UNSPSC Code:
12352200
€34.10
Please contact Customer Service for Availability
Request a Bulk OrderRequest a Sample
packaging
pkg of 12 ea
Quality Level
technique(s)
solid phase extraction (SPE): suitable
application(s)
environmental
food and beverages
compatibility
configured for plugging luer holes
General description
Male Luer plug is used to seal the inlet sampling cap which serves as an accessory for thermal desorption system storage containers.
Choose from one of the most recent versions:
Certificates of Analysis (COA)