Skip to Content
Merck
All Photos(1)

Key Documents

501298

Supelco

Carbon dioxide (99.8%)

cylinder of 48 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CO2
Molecular Weight:
44.01
EC Number:
MDL number:
UNSPSC Code:
23151816
PubChem Substance ID:

grade

analytical standard

Assay

99.8%

form

gas

packaging

cylinder of 48 L

manufacturer/tradename

Scott Gas T105-48

availability

not available in EU

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

SMILES string

O=C=O

InChI

1S/CO2/c2-1-3

InChI key

CURLTUGMZLYLDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

regulator

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Piercy et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 185(8), 891-903 (2015-08-20)
The present study determined whether EEG and/or EMG recordings could be used to reliably define activity states in the Brazilian black and white tegu lizard (Tupinambis merianae) and then examined the interactive effects of temperature and activity states on strategies
Erik H J Hulzebos et al.
Medicine and science in sports and exercise, 46(11), 2047-2052 (2014-05-23)
Lung function, nutritional status, and parameters of exercise capacity are known predictors of mortality in patients with cystic fibrosis (CF). The aim of the current study was to use these important parameters to develop a multivariate model to predict mortality
L Rienzi et al.
Reproductive biomedicine online, 30(1), 57-66 (2014-12-03)
Recent studies involving a limited number of patients have indicated a correlation between aneuploidy and various morphokinetic parameters during preimplantation development. The results among different groups, however, have been inconsistent in identifying the parameters that are able to predict chromosomal
Anne E Holland et al.
The European respiratory journal, 44(6), 1428-1446 (2014-11-02)
Field walking tests are commonly employed to evaluate exercise capacity, assess prognosis and evaluate treatment response in chronic respiratory diseases. In recent years, there has been a wealth of new literature pertinent to the conduct of the 6-min walk test
Xiaoqing Lu et al.
Blood, 125(4), 649-657 (2014-11-09)
Human germinal center-associated lymphoma (HGAL) is specifically expressed only in germinal center (GC) B lymphocytes and GC-derived lymphomas. HGAL protein decreases lymphocyte motility by inhibiting the ability of myosin to translocate actin via direct interaction with F-actin and myosin II

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service