Skip to Content
Merck
All Photos(1)

Documents

4S6831-U

Supelco

EPA 8260 Calibration Mix 2

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Agency

EPA 8260
610

CofA

current certificate can be downloaded

packaging

ampule of 1.5 mL

concentration

2000 μg/mL each component in methanol: water (19:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Recommended products

This product is currently being deleted and is being replaced by product 46831-U

Analyte

Description

    Acetone
    2-Butanone
    Carbon disulfide
    2-Chloroethyl vinyl ether
    2-Hexanone
    Iodomethane
    4-Methyl-2-pentanone
    Vinyl acetate
    See All (8)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system, Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service