Skip to Content
Merck
All Photos(1)

Key Documents

48045

Supelco

EPA 551A Halogenated Volatiles Mix

certified reference material, 2000 μg/mL each component in acetone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 551,551.1

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

EPA 551A halogenated volatiles mix has been used as a standard in the quenching of disinfection by-products in water like trihalomethanes and haloacetic acid by hydrogen peroxide as disinfectant using gas chromatography method of analysis (GC). It may be used as a certified reference standard for the quantification of trihalomethanes in environmental matrices using gas chromatography technique.[1]
EPA 551A halogenated volatiles mix has been used as a standard in the quenching of disinfection by-products in water like trihalomethanes and haloacetic acid by hydrogen peroxide as disinfectant using gas chromatography method of analysis (GC).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Bromodichloromethane
    Bromoform
    Carbon tetrachloride
    Chloroform
    Dibromochloromethane
    1,2-Dibromo-3-chloropropane
    1,2-Dibromoethane
    Tetrachloroethylene
    1,1,1-Trichloroethane
    Trichloroethylene
    See All (10)

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Ozone 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Supplementary Hazards

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quenching of chlorination disinfection by-product formation in drinking water by hydrogen peroxide
Batterman S, et al.
Water Research, 34, 1652-1658 (2000)
Analysis of trihalomethanes in water and air from indoor swimming pools using HS-SPME/GC/ECD
SA.A.S C, et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 46(4), 355-363 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service