Skip to Content
Merck
All Photos(1)

Key Documents

47787

Supelco

EPA 552.2 Acids Calibration Mix

certified reference material, in methyl tert-butyl ether (varied)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€88.50

€88.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€88.50

About This Item

UNSPSC Code:
12000000

€88.50


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Agency

EPA 552.2

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in methyl tert-butyl ether (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Bromoacetic acid 400 μg/mL
    Bromochloroacetic acid 400 μg/mL
    Bromdichloroacetic acid 400 μg/mL
    Chloroacetic acid 600 μg/mL
    Dibromochloroacetic acid 1000 μg/mL
    Dibromoacetic acid 200 μg/mL
    Dichloroacetic acid 600 μg/mL
    Tribromoacetic acid 2000 μg/mL
    Trichloroacetic acid 200 μg/mL
    See All (9)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-27.4 °F - closed cup

Flash Point(C)

-33.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service