Skip to Content
Merck
All Photos(1)

Key Documents

47991-U

Supelco

EPA TCL Base-Neutrals Mix

certified reference material, 2000 μg/mL each component in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA
610

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

Certified reference material (CRM) standards such as EPA TCL Base-Neutrals Mix are specifically designed for monitoring organic chemicals on the US EPA′s Target Compound List (TCL) in water, sediment, or soil from hazardous waste sites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Azobenzene
    Benzyl butyl phthalate
    Bis(2-chloroethoxy)methane
    Bis(2-chloroethyl) ether
    Bis(2-ethylhexyl) phthalate
    4-Bromodiphenyl ether
    Carbazole
    4-Chlorodiphenyl ether
    Bis-(2-chloroisopropyl) ether
    2-Chloronaphthalene
    Dibutyl phthalate
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Diethyl phthalate
    Dimethyl phthalate
    2,4-Dinitrotoluene
    2,6-Dinitrotoluene
    Di-n-octyl phthalate
    Hexachlorobenzene
    Hexachloro-1,3-butadiene
    Hexachlorocyclopentadiene
    Hexachloroethane
    Isophorone
    Nitrobenzene
    N-Nitrosodimethylamine
    N-Nitrosodiphenylamine
    N-Nitrosodi-n-propylamine
    1,2,4-Trichlorobenzene
    See All (29)

Signal Word

Danger

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

EU REACH Annex XIV (Authorisation List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service