Skip to Content
Merck
All Photos(1)

Key Documents

506559

Supelco

EPA 625 Semivolatile Calibration Mix

1000 μg/mL each component in methylene chloride: benzene (3:1), analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€135.00

€135.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€135.00

About This Item

UNSPSC Code:
12000000

€135.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Agency

EPA 625

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in methylene chloride: benzene (3:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Anthracene
    Azobenzene
    Benz[a]anthracene
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzo[a]pyrene
    Benzyl butyl phthalate
    Bis(2-chloroethoxy)methane
    Bis(2-chloroethyl) ether
    Bis(2-ethylhexyl) phthalate
    4-Bromodiphenyl ether
    Carbazole
    4-Chlorodiphenyl ether
    Bis-(2-chloroisopropyl) ether
    4-Chloro-3-methylphenol
    2-Chloronaphthalene
    2-Chlorophenol
    Chrysene
    Dibenz[a,h]anthracene
    Dibutyl phthalate
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    2,4-Dichlorophenol
    Diethyl phthalate
    2,4-Dimethylphenol
    Dimethyl phthalate
    2,4-Dinitrophenol
    2,4-Dinitrotoluene
    2,6-Dinitrotoluene
    Di-n-octyl phthalate
    Fluoranthene
    Fluorene
    Hexachlorobenzene
    Hexachloro-1,3-butadiene
    Hexachlorocyclopentadiene
    Hexachloroethane
    Indeno[1,2,3-cd]pyrene
    Isophorone
    2-Methyl-4,6-dinitrophenol
    Naphthalene
    Nitrobenzene
    2-Nitrophenol
    4-Nitrophenol
    N-Nitrosodimethylamine
    N-Nitrosodi-n-propylamine
    Pentachlorophenol
    Phenanthrene
    Phenol
    Pyrene
    1,2,4-Trichlorobenzene
    2,4,6-Trichlorophenol
    See All (55)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Blood, Central nervous system, Respiratory system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service