Skip to Content
Merck
All Photos(1)

Key Documents

47827

Supelco

Sodium Cyclamate

analytical standard

Synonym(s):

Sodium N-cyclohexylsulfamate, N-Cyclohexylsulfamic acid sodium salt, Cyclamic acid sodium salt, Cyclohexanesulfamic acid sodium salt, Sodium cyclamate, Sodium cyclohexanesulfamate

Sign Into View Organizational & Contract Pricing

Select a Size

1000 MG
€37.80

€37.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1000 MG
€37.80

About This Item

Linear Formula:
C6H11NHSO3Na
CAS Number:
Molecular Weight:
201.22
Beilstein:
4166868
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E952

€37.80


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

[Na+].[O-]S(=O)(=O)NC1CCCCC1

InChI

1S/C6H13NO3S.Na/c8-11(9,10)7-6-4-2-1-3-5-6;/h6-7H,1-5H2,(H,8,9,10);/q;+1/p-1

InChI key

UDIPTWFVPPPURJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sodium cyclamate may be used as a reference standard in the determination of sodium cyclamate in artificial food and beverages using ion chromatography (IC).[1]

Biochem/physiol Actions

Phenylsulfamate sweet tastant detectable by humans. ′First generation′ sweetener studied in animal models for its carcinogenic potential.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation and simultaneous determination of four artificial sweeteners in food and beverages by ion chromatography.
Zhu Y, et al.
Journal of Chromatography A, 1085(1), 143-146 (2005)
Miesje van der Stoel et al.
Journal of cell science, 133(3) (2020-01-23)
Endothelial YAP/TAZ (YAP is also known as YAP1, and TAZ as WWTR1) signaling is crucial for sprouting angiogenesis and vascular homeostasis. However, the underlying molecular mechanisms that explain how YAP/TAZ control the vasculature remain unclear. This study reveals that the

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service