Skip to Content
Merck
All Photos(1)

Key Documents

80054

Supelco

Advantame®

analytical standard

Synonym(s):

N-[N-[3-(3-Hydroxy-4-methoxyphenyl)propyl]-α-L-aspartyl]-L-phenylalanine 1-methyl ester monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€62.40

€62.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€62.40

About This Item

Empirical Formula (Hill Notation):
C24H30N2O7 · H2O
CAS Number:
Molecular Weight:
476.52
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

€62.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

2.5-5.0% water

application(s)

food and beverages
personal care

format

neat

SMILES string

O.COC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CC(O)=O)NCCCc2ccc(OC)c(O)c2

InChI

1S/C24H30N2O7.H2O/c1-32-21-11-10-17(14-20(21)27)9-6-12-25-18(15-22(28)29)23(30)26-19(24(31)33-2)13-16-7-4-3-5-8-16;/h3-5,7-8,10-11,14,18-19,25,27H,6,9,12-13,15H2,1-2H3,(H,26,30)(H,28,29);1H2/t18-,19-;/m0./s1

InChI key

NQQIEOGMDRFTDZ-HLRBRJAUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Advantame® is a high intensity, non-caloric sweetener, which is derived from aspartame.[1][2] It is widely used in food and beverage industry.[1][2]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

Advantame is a registered trademark of Ajinomoto Co., Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Acute and multiple-dose studies to determine the safety, tolerability, and pharmacokinetic profile of advantame in healthy volunteers.
Warrington S, et al.
Food And Chemical Toxicology, 49, S77-S83 (2011)
Advantame sweetener preference in C57BL/6J mice and Sprague-Dawley rats.
Sclafani A and Ackroff K
Chemical Senses, 40(3), 181-186 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service