Skip to Content
Merck
All Photos(1)

Key Documents

46831-U

Supelco

EPA 8260 Calibration Mix 2

analytical standard, 2000 μg/mL each component in methanol: water (19:1)

Sign Into View Organizational & Contract Pricing

Select a Size

1.5 ML
€48.00

€48.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1.5 ML
€48.00

About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

€48.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Agency

EPA 8260

description

Separate Source

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 × 1.5 mL (46831-U)

concentration

2000 μg/mL each component in methanol: water (19:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

General description

EPA 8260 Calibration Mix 2 is a multi-component solution containing analytes such as acetone, 2-butanone, carbon disulfide, 2-chloroethyl vinyl ether, 2-hexanone, iodomethane, 4-methyl-2-pentanone, and vinyl acetate. It may be used for identifying and quantifying volatile organic compounds (VOCs) on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. The pre-requisite of EPA 8260 method is the use of gas chromatography (GC) with a selective detector or gas chromatography/mass spectrometry (GC/MS) or high-performance liquid chromatography (HPLC).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acetone
    2-Butanone
    Carbon disulfide
    2-Chloroethyl vinyl ether
    2-Hexanone
    Iodomethane
    4-Methyl-2-pentanone
    Vinyl acetate
    See All (8)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system, Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service