Skip to Content
Merck
All Photos(1)

Key Documents

47428-U

Supelco

EPA 524.2 Rev 4 Update Ketones Mix

certified reference material, 2000 μg/mL in methanol: water (9:1)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€41.70

€41.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€41.70

About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

€41.70


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 524.2

description

Separate Source

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 × 1 mL (47428-U)

concentration

2000 μg/mL in methanol: water (9:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

EPA 524.2 Rev 4 Update Ketones Mix is a multi-component solution containing acetone, 2-butanone, 1,1-dichloro-2-propanone, 2-hexanone, and 4-methyl-2-pentanone as certified reference materials (CRMs).
US EPA 500 Series methods are analytical methods for identifying and quantifying organic contaminants such as volatile organic compounds (VOCs), pesticides, synthetic organic compounds (SOCs), and trihalomethane disinfection byproducts (THMs) in raw source water, finished drinking water, and drinking water at all stages of treatment.

Application

EPA 524.2 Rev 4 Update Ketones Mix may be used as a CRM to quantify volatile organic compound (VOC) analytes in indoor and outdoor air samples by gas chromatography-mass spectrometry (GC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acetone
    2-Butanone
    1,1-Dichloro-2-propanone
    2-Hexanone
    4-Methyl-2-pentanone

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile organic compounds (VOCs) in conventional and high performance school buildings in the US
Zhong L, et al.
International Journal of Environmental Research and Public Health, 14(1), 100-100 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service