Skip to Content
Merck
All Photos(1)

Key Documents

46401-U

Supelco

rac-β-Tocopherol solution

50 mg/mL in hexane, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
85151701
eCl@ss:
34058016
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

50 mg/mL in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

single component solution

storage temp.

-10 to -25°C

SMILES string

O1[C@@](CCc2c1c(cc(c2C)O)C)(CCCC(CCCC(CCCC(C)C)C)C)C

InChI

1S/C28H48O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-24(6)26(29)19-23(5)27(25)30-28/h19-22,29H,8-18H2,1-7H3/t21?,22?,28-/m1/s1

InChI key

WGVKWNUPNGFDFJ-CBIUGAAKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Tocopherol is a form of fat-soluble vitamin E and shows anti-oxidant, anti-inflammatory, anti-cell proliferative, and anti-cancer properties.

Application

The analytical standard in solution can also be used as follows:
  • Simultaneous determination of vitamin A and all four tocopherol isomers in human serum samples by ultra-high performance liquid chromatography (UHPLC) combined with fluorescence detection following their liquid-liquid extraction (LLE)
  • Multi-residue analysis of vitamin B (B1, B2, B5, and B9), and vitamin E (α-tocopherol, β-tocopherol, α-tocotrienol, and β-tocotrienol) in grass and grain samples of four different wheat varieties by liquid chromatography electrospray ionization/multistage mass spectrometry (LC-ESI-MS/MS)
  • Separation and estimation of four tocopherols and four tocotrienols in pecan nut cultivar samples using non-aqueous reversed-phase high-performance liquid chromatography (NARP-HPLC) coupled with atmospheric pressure chemical ionization-tandem mass spectrometric (APCI-MS/MS) detection
  • Analysis of different cold pressed oil samples to detect and quantify eight tocol homologs using a chemometrically-assisted optimized high-performance liquid chromatography (HPLC) with fluorescence detection (FLD) based method
  • Development and validation of normal-phase high-performance liquid chromatography (NP-HPLC) method, along with fluorescence detection, to simultaneously determine four tocopherols and four lignans in sesame oil samples

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pressurized liquid extraction and dispersive liquid-liquid microextraction for determination of tocopherols and tocotrienols in plant foods by liquid chromatography with fluorescence and atmospheric pressure chemical ionization-mass spectrometry detection
Vi?as P, et al.
Talanta, 119(9), 98-104 (2014)
Rapid determination of lipophilic vitamins in human serum by ultra-high performance liquid chromatography using a fluorinated column and high-throughput miniaturized liquid-liquid extraction
Cervinkova B, et al.
Journal of Separation Science, 40(17), 3375-3382 (2017)
Qualitative and quantitative analysis of retinol, retinyl esters, tocopherols and selected carotenoids out of various internal organs from different species by HPLC
Schaffer MW, et al.
Analytical Methods : Advancing Methods and Applications, 2(9), 1320-1332 (2010)
Analysis of the components of pumpkin seed oil in suppositories and the possibility of its use in pharmaceuticals
A.O.V, et al.
Journal of Chemical and Pharmaceutical Research , 6(5), 1106-1116 (2014)
Xiaofeng Fang et al.
Developmental cell, 48(3), 371-382 (2019-01-01)
As integral regulators in plant development and stress response, microRNAs (miRNAs) themselves need to be tightly regulated. Here, we show that tocopherols (vitamin E), lipid-soluble antioxidants synthesized from tyrosine in chloroplasts, positively regulate the biogenesis of miRNAs. Tocopherols are required

Protocols

GB method with Ascentis® Express C30 column achieves baseline resolution for Vitamin A and E critical pair.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service