V-020
(±)−α−Tocopherol (Vitamin E) solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
(±)-α-Tocopherol, DL-all-rac-α-Tocopherol, Vitamin E
About This Item
Recommended Products
grade
certified reference material
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1.0 mg/mL in methanol
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
density
0.950 g/mL at 20 °C (lit.)
application(s)
clinical testing
clinical testing
format
single component solution
storage temp.
−70°C
SMILES string
CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1
InChI
1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3
InChI key
GVJHHUAWPYXKBD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Simultaneous determination of vitamin A and all four tocopherol isomers in human serum samples by ultra-high performance liquid chromatography (UHPLC) combined with fluorescence detection following their liquid-liquid extraction (LLE)
- Multi-residue analysis of vitamin B (B1, B2, B5, and B9), and vitamin E (α-tocopherol, β-tocopherol, α-tocotrienol, and β-tocotrienol) in grass and grain samples of four different wheat varieties by liquid chromatography-electrospray ionization/multistage mass spectrometry (LC-ESI-MS/MS)
- Separation and estimation of four tocopherols and four tocotrienols in pecan nut cultivar samples using non-aqueous reversed-phase high-performance liquid chromatography (NARP-HPLC) coupled with atmospheric pressure chemical ionization-tandem mass spectrometric (APCI-MS/MS) detection
- Analysis of different cold pressed oil samples to detect and quantify eight tocol- homologs using a chemometrically-assisted optimized high-performance liquid chromatography (HPLC) with fluorescence detection (FLD) based method
- High-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) based combined analysis of retinol, α‐tocopherol, 25‐hydroxyvitamin D2, and 25‐hydroxyvitamin in plasma samples of cardiovascular patients
Features and Benefits
- Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
- Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
- Rigorously tested through real-time stability studies to ensure accuracy and shelf life
- Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
- Flame sealed under argon into ampoules for long-term shelf life
- Offered in a convenient, DEA-exempt format to improve laboratory efficiency
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
HPLC separates vitamin E acetate, THC, and CBD in vape products, providing accurate analysis of commercially available vape liquids.
Protocols
Separation of (±)-α-Tocopherol, analytical standard; Menadione (K3), analytical standard; (+)-γ-Tocopherol, analytical standard; Cholecalciferol (D3), analytical standard
Separation of δ-Tocopherol, analytical standard; Retinyl acetate; (+)-γ-Tocopherol; α-Tocopherol, ≥95.5%; Retinol; Vitamin A (acetate), meets USP testing specifications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service