Skip to Content
Merck
All Photos(2)

Documents

44896-U

Supelco

ASTM® D6584 1,2,4-Butanetriol Solution

greener alternative

1000 μg/mL in pyridine, analytical standard

Synonym(s):

Biodiesel impurity, Butanetriol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Agency

ASTM® D6584

packaging

pkg of 5 mL

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

concentration

1000 μg/mL in pyridine

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

single component solution

greener alternative category

storage temp.

2-8°C

InChI

1S/C4H10O3/c5-2-1-4(7)3-6/h4-7H,1-3H2/t4-/m0/s1

InChI key

ARXKVVRQIIOZGF-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product is a Biobased products, showing key improvements in Green Chemistry Principles “Safer Solvents and Auxiliaries".

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service